Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 20 - 21 | 4105 | 92 | |
| XM_015769940.2 | XP_015625426.1 | 19 - 20 | 3868 | 92 | ||
| GenBank mRNA | AB477099.1 | - | 19 - 20 | 3791 | 92 | |
| AK064381.1 | - | 1 - 2 | 216 | 92 | ||
| AK121261.1 | - | 20 - 21 | 4105 | 92 | ||
| AK121875.1 | - | 20 - 21 | 4060 | 92 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map