Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 2 - 3 | 431 | 71 | |
| XM_015769940.2 | XP_015625426.1 | 1 - 2 | 194 | 71 | ||
| GenBank mRNA | AB477099.1 | - | 1 - 2 | 117 | 71 | |
| AK121261.1 | - | 2 - 3 | 431 | 71 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map