Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 25 - 26 | 5504 | 80 | |
| XM_015769940.2 | XP_015625426.1 | 24 - 25 | 5267 | 80 | ||
| GenBank mRNA | AB477099.1 | - | 24 - 25 | 5190 | 80 | |
| AK064381.1 | - | 6 - 7 | 1615 | 80 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map