Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 26 - 27 | 5614 | 77 | |
| XM_015769940.2 | XP_015625426.1 | 25 - 26 | 5377 | 77 | ||
| GenBank mRNA | AB477099.1 | - | 25 - 26 | 5300 | 77 | |
| AK064381.1 | - | 7 - 8 | 1725 | 77 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map