Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 27 - 28 | 5742 | 147 | |
| XM_015769940.2 | XP_015625426.1 | 26 - 27 | 5505 | 147 | ||
| GenBank mRNA | AB477099.1 | - | 26 - 27 | 5428 | 147 | |
| AK064381.1 | - | 8 - 9 | 1853 | 147 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map