Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 30 - 31 | 6499 | 101 | |
| XM_015769940.2 | XP_015625426.1 | 29 - 30 | 6262 | 101 | ||
| GenBank mRNA | AB477099.1 | - | 29 - 30 | 6185 | 101 | |
| AK064381.1 | - | 11 - 12 | 2610 | 101 | ||
| AK072218.1 | - | 4 - 5 | 784 | 101 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map