Assay Details
| Gene Symbol: LOC4330484 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769939.2 | XP_015625425.1 | 31 - 32 | 6928 | 61 | |
| XM_015769940.2 | XP_015625426.1 | 30 - 31 | 6691 | 61 | ||
| GenBank mRNA | AK064381.1 | - | 12 - 13 | 3039 | 61 | |
| AK072218.1 | - | 5 - 6 | 1213 | 61 | ||
More Information
Set Membership: |
|

LOC4330484
Genomic Map