Assay Details
| Gene Symbol: LOC4330587 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769290.2 | XP_015624776.1 | 1 - 2 | 82 | 87 | |
| GenBank mRNA | AJ300218.1 | - | 1 - 2 | 75 | 87 | |
| AK241920.1 | - | 1 - 2 | 84 | 87 | ||
| EU847013.1 | - | 1 - 2 | 66 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AJ300218.1, XM_015769290.2 transcripts.
Set Membership: |
|

LOC4330587
Genomic Map