Assay Details
| Gene Symbol: LOC4330726 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771971.2 | XP_015627457.1 | 3 - 4 | 616 | 125 | |
| XM_015771972.2 | XP_015627458.1 | 3 - 4 | 607 | 125 | ||
| XM_015771973.2 | XP_015627459.1 | 4 - 5 | 654 | 125 | ||
| XM_015771975.2 | XP_015627461.1 | 4 - 5 | 651 | 125 | ||
| XM_015771976.2 | XP_015627462.1 | 4 - 5 | 357 | 125 | ||
| XM_015771977.2 | XP_015627463.1 | 3 - 4 | 176 | 125 | ||
| GenBank mRNA | AK101365.1 | - | 3 - 4 | 581 | 125 | |
More Information
Set Membership: |
|

LOC4330726
Genomic Map