Assay Details
| Gene Symbol: LOC4330909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770404.2 | XP_015625890.1 | 12 - 13 | 1256 | 102 | |
| XM_015770405.2 | XP_015625891.1 | 12 - 13 | 1177 | 102 | ||
| XM_015770406.2 | XP_015625892.1 | 12 - 13 | 1084 | 102 | ||
| XM_015770407.2 | XP_015625893.1 | 13 - 14 | 981 | 102 | ||
| XM_015770408.2 | XP_015625894.1 | 11 - 12 | 1360 | 102 | ||
| GenBank mRNA | AK058466.1 | - | 12 - 13 | 1219 | 102 | |
| AK100242.1 | - | 11 - 12 | 1751 | 102 | ||
More Information
Set Membership: |
|

LOC4330909
Genomic Map