Assay Details
| Gene Symbol: LOC4330909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770404.2 | XP_015625890.1 | 1 - 2 | 418 | 107 | |
| GenBank mRNA | AK058466.1 | - | 1 - 2 | 381 | 107 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770404.2 transcripts.
Set Membership: |
|

LOC4330909
Genomic Map