Assay Details
| Gene Symbol: LOC4330909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770404.2 | XP_015625890.1 | 2 - 3 | 505 | 114 | |
| XM_015770405.2 | XP_015625891.1 | 2 - 3 | 426 | 114 | ||
| XM_015770406.2 | XP_015625892.1 | 2 - 3 | 333 | 114 | ||
| XM_015770407.2 | XP_015625893.1 | 3 - 4 | 230 | 114 | ||
| GenBank mRNA | AK058466.1 | - | 2 - 3 | 468 | 114 | |
| AK100242.1 | - | 1 - 2 | 1000 | 114 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015770406.2, XM_015770407.2 transcripts.
Set Membership: |
|

LOC4330909
Genomic Map