Assay Details
| Gene Symbol: LOC4330909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770404.2 | XP_015625890.1 | 3 - 4 | 613 | 72 | |
| XM_015770405.2 | XP_015625891.1 | 3 - 4 | 534 | 72 | ||
| XM_015770406.2 | XP_015625892.1 | 3 - 4 | 441 | 72 | ||
| XM_015770407.2 | XP_015625893.1 | 4 - 5 | 338 | 72 | ||
| XM_015770408.2 | XP_015625894.1 | 2 - 3 | 717 | 72 | ||
| GenBank mRNA | AK058466.1 | - | 3 - 4 | 576 | 72 | |
| AK100242.1 | - | 2 - 3 | 1108 | 72 | ||
More Information
Set Membership: |
|

LOC4330909
Genomic Map