Assay Details
| Gene Symbol: LOC4330909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770404.2 | XP_015625890.1 | 6 - 7 | 802 | 81 | |
| XM_015770405.2 | XP_015625891.1 | 6 - 7 | 723 | 81 | ||
| XM_015770406.2 | XP_015625892.1 | 6 - 7 | 630 | 81 | ||
| XM_015770407.2 | XP_015625893.1 | 7 - 8 | 527 | 81 | ||
| XM_015770408.2 | XP_015625894.1 | 5 - 6 | 906 | 81 | ||
| GenBank mRNA | AK058466.1 | - | 6 - 7 | 765 | 81 | |
| AK100242.1 | - | 5 - 6 | 1297 | 81 | ||
More Information
Set Membership: |
|

LOC4330909
Genomic Map