Assay Details
| Gene Symbol: LOC4330909 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015770404.2 | XP_015625890.1 | 9 - 10 | 1049 | 98 | |
| XM_015770405.2 | XP_015625891.1 | 9 - 10 | 970 | 98 | ||
| XM_015770406.2 | XP_015625892.1 | 9 - 10 | 877 | 98 | ||
| XM_015770407.2 | XP_015625893.1 | 10 - 11 | 774 | 98 | ||
| XM_015770408.2 | XP_015625894.1 | 8 - 9 | 1153 | 98 | ||
| GenBank mRNA | AK058466.1 | - | 9 - 10 | 1012 | 98 | |
| AK100242.1 | - | 8 - 9 | 1544 | 98 | ||
More Information
Set Membership: |
|

LOC4330909
Genomic Map