Assay Details
| Gene Symbol: LOC4330925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768702.2 | XP_015624188.1 | 2 - 3 | 451 | 113 | |
| GenBank mRNA | AK060474.1 | - | 2 - 3 | 459 | 113 | |
| AK073762.1 | - | 2 - 3 | 458 | 113 | ||
More Information
Set Membership: |
|

LOC4330925
Genomic Map