Assay Details
| Gene Symbol: LOC4330925 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768702.2 | XP_015624188.1 | 3 - 4 | 614 | 80 | |
| GenBank mRNA | AK060474.1 | - | 3 - 4 | 622 | 80 | |
| AK073762.1 | - | 3 - 4 | 621 | 80 | ||
| AK119213.1 | - | 2 - 3 | 440 | 80 | ||
More Information
Set Membership: |
|

LOC4330925
Genomic Map