Assay Details
| Gene Symbol: LOC4330930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771474.2 | XP_015626960.1 | 6 - 7 | 1867 | 87 | |
| GenBank mRNA | AK065421.1 | - | 6 - 7 | 1400 | 87 | |
More Information
Set Membership: |
|

LOC4330930
Genomic Map