Assay Details
| Gene Symbol: LOC4331050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769665.2 | XP_015625151.1 | 4 - 5 | 629 | 111 | |
| GenBank mRNA | AB037155.1 | - | 4 - 5 | 580 | 111 | |
| AK062185.1 | - | 4 - 5 | 596 | 111 | ||
| AK067051.1 | - | 4 - 5 | 496 | 111 | ||
| AK101474.1 | - | 4 - 5 | 623 | 111 | ||
| AK103936.1 | - | 4 - 5 | 609 | 111 | ||
| D17788.1 | - | 4 - 5 | 579 | 111 | ||
More Information
Set Membership: |
|

LOC4331050
Genomic Map