Assay Details
| Gene Symbol: LOC4331050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769665.2 | XP_015625151.1 | 6 - 7 | 938 | 119 | |
| GenBank mRNA | AB037155.1 | - | 6 - 7 | 889 | 119 | |
| AK062185.1 | - | 6 - 7 | 905 | 119 | ||
| AK067051.1 | - | 6 - 7 | 805 | 119 | ||
| AK101474.1 | - | 6 - 7 | 932 | 119 | ||
| AK103936.1 | - | 6 - 7 | 918 | 119 | ||
| D17788.1 | - | 6 - 7 | 888 | 119 | ||
More Information
Set Membership: |
|

LOC4331050
Genomic Map