Assay Details
| Gene Symbol: LOC4331050 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769665.2 | XP_015625151.1 | 9 - 10 | 1294 | 67 | |
| GenBank mRNA | AB037155.1 | - | 9 - 10 | 1245 | 67 | |
| AK062185.1 | - | 9 - 10 | 1261 | 67 | ||
| AK067051.1 | - | 9 - 10 | 1161 | 67 | ||
| AK101474.1 | - | 9 - 10 | 1288 | 67 | ||
| AK103936.1 | - | 9 - 10 | 1274 | 67 | ||
| D17788.1 | - | 9 - 10 | 1244 | 67 | ||
More Information
Set Membership: |
|

LOC4331050
Genomic Map