Assay Details
| Gene Symbol: LOC4331058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771350.2 | XP_015626836.1 | 1 - 2 | 447 | 62 | |
| GenBank mRNA | AK068801.1 | - | 1 - 2 | 339 | 62 | |
More Information
Set Membership: |
|

LOC4331058
Genomic Map