Assay Details
| Gene Symbol: LOC4331058 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015771350.2 | XP_015626836.1 | 7 - 8 | 1232 | 109 | |
| GenBank mRNA | AK068801.1 | - | 7 - 8 | 1124 | 109 | |
More Information
Set Membership: |
|

LOC4331058
Genomic Map