Assay Details
| Gene Symbol: LOC4331098 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015768292.2 | XP_015623778.1 | 2 - 3 | 1113 | 78 | |
| GenBank mRNA | AK060976.1 | - | 2 - 3 | 593 | 78 | |
| AK070982.1 | - | 2 - 3 | 720 | 78 | ||
More Information
Set Membership: |
|

LOC4331098
Genomic Map