Assay Details
| Gene Symbol: LOC4331151 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015769947.2 | XP_015625433.1 | 1 - 2 | 193 | 65 | |
| GenBank mRNA | AK068473.1 | - | 1 - 2 | 140 | 65 | |
| AK100685.1 | - | 1 - 2 | 139 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015769947.2 transcripts.
Set Membership: |
|

LOC4331151
Genomic Map