Assay Details
| Gene Symbol: LOC4331345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777223.1 | XP_015632709.1 | 14 - 15 | 2071 | 99 | |
| GenBank mRNA | AJ556802.1 | - | 4 - 5 | 536 | 99 | |
| AK059498.1 | - | 3 - 4 | 402 | 99 | ||
| AK102378.1 | - | 14 - 15 | 2065 | 99 | ||
| AY425991.1 | - | 14 - 15 | 2065 | 99 | ||
More Information
Set Membership: |
|

LOC4331345
Genomic Map