Assay Details
| Gene Symbol: LOC4331345 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777223.1 | XP_015632709.1 | 16 - 17 | 2530 | 101 | |
| GenBank mRNA | AK059498.1 | - | 5 - 6 | 861 | 101 | |
| AK102378.1 | - | 16 - 17 | 2524 | 101 | ||
| AY425991.1 | - | 16 - 17 | 2524 | 101 | ||
More Information
Set Membership: |
|

LOC4331345
Genomic Map