Assay Details
| Gene Symbol: LOC4331492 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776005.2 | XP_015631491.1 | 2 - 3 | 521 | 82 | |
| XM_015776006.2 | XP_015631492.1 | 2 - 3 | 518 | 82 | ||
| XM_015776007.2 | XP_015631493.1 | 2 - 3 | 456 | 82 | ||
| XM_015776008.2 | XP_015631494.1 | 2 - 3 | 518 | 82 | ||
| XM_015776009.2 | XP_015631495.1 | 2 - 3 | 515 | 82 | ||
| GenBank mRNA | AK073895.1 | - | 2 - 3 | 432 | 82 | |
| DQ838016.1 | - | 2 - 3 | 401 | 82 | ||
More Information
Set Membership: |
|

LOC4331492
Genomic Map