Assay Details
| Gene Symbol: LOC4331511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774260.2 | XP_015629746.1 | 1 - 2 | 192 | 134 | |
| GenBank mRNA | AK103714.1 | - | 1 - 2 | 129 | 134 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015774260.2 transcripts.
Set Membership: |
|

LOC4331511
Genomic Map