Assay Details
| Gene Symbol: LOC4331511 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015774260.2 | XP_015629746.1 | 7 - 8 | 779 | 144 | |
| GenBank mRNA | AK103714.1 | - | 7 - 8 | 716 | 144 | |
More Information
Set Membership: |
|

LOC4331511
Genomic Map