Assay Details
| Gene Symbol: LOC4331634 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775810.2 | XP_015631296.1 | 1 | 1685 | 82 | |
| GenBank mRNA | AK106694.1 | - | 1 | 1652 | 82 | |
More Information
Set Membership: |
|

LOC4331634
Genomic Map