Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 10 - 11 | 1405 | 65 | |
| GenBank mRNA | AF499737.1 | - | 10 - 11 | 1113 | 65 | |
| AF508799.1 | - | 10 - 11 | 1119 | 65 | ||
| AK102192.1 | - | 10 - 11 | 1404 | 65 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map