Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 11 - 12 | 1566 | 81 | |
| GenBank mRNA | AF499737.1 | - | 11 - 12 | 1274 | 81 | |
| AF508799.1 | - | 11 - 12 | 1280 | 81 | ||
| AK102192.1 | - | 11 - 12 | 1565 | 81 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map