Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 12 - 13 | 1747 | 79 | |
| GenBank mRNA | AK060645.1 | - | 1 - 2 | 50 | 79 | |
| AK102192.1 | - | 12 - 13 | 1746 | 79 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map