Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 13 - 14 | 1857 | 63 | |
| GenBank mRNA | AF499737.1 | - | 13 - 14 | 1565 | 63 | |
| AF508799.1 | - | 13 - 14 | 1571 | 63 | ||
| AK060645.1 | - | 2 - 3 | 160 | 63 | ||
| AK102192.1 | - | 13 - 14 | 1856 | 63 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map