Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 14 - 15 | 1961 | 80 | |
| GenBank mRNA | AF499737.1 | - | 14 - 15 | 1669 | 80 | |
| AF508799.1 | - | 14 - 15 | 1675 | 80 | ||
| AK060645.1 | - | 3 - 4 | 264 | 80 | ||
| AK102192.1 | - | 14 - 15 | 1960 | 80 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map