Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 16 - 17 | 2187 | 78 | |
| GenBank mRNA | AF499737.1 | - | 16 - 17 | 1901 | 78 | |
| AF508799.1 | - | 16 - 17 | 1907 | 78 | ||
| AK060645.1 | - | 5 - 6 | 490 | 78 | ||
| AK102192.1 | - | 16 - 17 | 2186 | 78 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map