Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 18 - 19 | 2724 | 104 | |
| GenBank mRNA | AF499737.1 | - | 18 - 19 | 2438 | 104 | |
| AF508799.1 | - | 18 - 19 | 2444 | 104 | ||
| AK060645.1 | - | 7 - 8 | 1027 | 104 | ||
| AK102192.1 | - | 18 - 19 | 2723 | 104 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map