Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 2 - 3 | 578 | 70 | |
| GenBank mRNA | AF499737.1 | - | 2 - 3 | 286 | 70 | |
| AF508799.1 | - | 2 - 3 | 292 | 70 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map