Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 3 - 4 | 698 | 74 | |
| GenBank mRNA | AK102192.1 | - | 3 - 4 | 697 | 74 | |
More Information
Set Membership: |
|

LOC4331730
Genomic Map