Assay Details
| Gene Symbol: LOC4331730 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015777557.2 | XP_015633043.1 | 4 - 5 | 763 | 77 | |
| GenBank mRNA | AF499737.1 | - | 4 - 5 | 471 | 77 | |
| AF508799.1 | - | 4 - 5 | 477 | 77 | ||
| AK102192.1 | - | 4 - 5 | 762 | 77 | ||
More Information
Set Membership: |
|

LOC4331730
Genomic Map