Assay Details
| Gene Symbol: LOC4331975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776675.2 | XP_015632161.1 | 3 - 4 | 815 | 82 | |
| XM_015776676.2 | XP_015632162.1 | 3 - 4 | 815 | 82 | ||
| GenBank mRNA | AK065564.1 | - | 2 - 3 | 1013 | 82 | |
| AK067825.1 | - | 3 - 4 | 604 | 82 | ||
More Information
Set Membership: |
|

LOC4331975
Genomic Map