Assay Details
| Gene Symbol: LOC4331975 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015776676.2 | XP_015632162.1 | 5 - 6 | 1179 | 141 | |
| GenBank mRNA | AK065564.1 | - | 4 - 5 | 1377 | 141 | |
More Information
Set Membership: |
|

LOC4331975
Genomic Map