Assay Details
| Gene Symbol: LOC4332258 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772666.2 | XP_015628152.1 | 1 | 1935 | 141 | |
| GenBank mRNA | AK121154.1 | - | 1 | 1846 | 141 | |
More Information
Set Membership: |
|

LOC4332258
Genomic Map