Assay Details
| Gene Symbol: LOC4332425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775289.1 | XP_015630775.1 | 1 - 2 | 298 | 144 | |
| GenBank mRNA | AK103544.1 | - | 1 - 2 | 144 | 144 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015775289.1 transcripts.
Set Membership: |
|

LOC4332425
Genomic Map