Assay Details
| Gene Symbol: LOC4332425 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775287.2 | XP_015630773.1 | 3 - 4 | 1928 | 121 | |
| XM_015775289.1 | XP_015630775.1 | 2 - 3 | 1886 | 121 | ||
| XM_015775290.2 | XP_015630776.1 | 2 - 3 | 1739 | 121 | ||
| GenBank mRNA | AK064303.1 | - | 1 - 2 | 1123 | 121 | |
| AK103544.1 | - | 2 - 3 | 1732 | 121 | ||
More Information
Set Membership: |
|

LOC4332425
Genomic Map