Assay Details
| Gene Symbol: LOC4332513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015772344.2 | XP_015627830.1 | 1 - 2 | 351 | 110 | |
| GenBank mRNA | AK069970.1 | - | 1 - 2 | 243 | 110 | |
More Information
Set Membership: |
|

LOC4332513
Genomic Map