Assay Details
| Gene Symbol: LOC4332534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775586.2 | XP_015631072.1 | 17 - 18 | 1999 | 83 | |
| XM_015775587.2 | XP_015631073.1 | 17 - 18 | 2082 | 83 | ||
| XM_015775589.2 | XP_015631075.1 | 13 - 14 | 1445 | 83 | ||
| GenBank mRNA | AK067856.1 | - | 17 - 18 | 1961 | 83 | |
More Information
Set Membership: |
|

LOC4332534
Genomic Map