Assay Details
| Gene Symbol: LOC4332534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775586.2 | XP_015631072.1 | 18 - 19 | 2241 | 95 | |
| XM_015775587.2 | XP_015631073.1 | 18 - 19 | 2324 | 95 | ||
| XM_015775589.2 | XP_015631075.1 | 14 - 15 | 1687 | 95 | ||
| GenBank mRNA | AK067856.1 | - | 18 - 19 | 2203 | 95 | |
More Information
Set Membership: |
|

LOC4332534
Genomic Map