Assay Details
| Gene Symbol: LOC4332534 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015775586.2 | XP_015631072.1 | 8 - 9 | 1110 | 111 | |
| XM_015775587.2 | XP_015631073.1 | 8 - 9 | 1196 | 111 | ||
| XM_015775589.2 | XP_015631075.1 | 4 - 5 | 556 | 111 | ||
| GenBank mRNA | AK067856.1 | - | 8 - 9 | 1072 | 111 | |
More Information
Set Membership: |
|

LOC4332534
Genomic Map